Blue Prestained Protein Standard, Broad Range (11 to 250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Certificates
About this item
The Blue prestained protein standard, Broad range is a mixture of highly pure, recombinant, prestained proteins, covalently coupled with a blue chromophore, that resolves into 11 sharp bands when electrophoresed.
- Allows approximate molecular weight determination when performing SDS-PAGE analysis
- Applications include verification of Western transfer efficiency and fluorescent imaging of SDS-PAGE
- Direct loading, additional loading buffer and heat incubation not required
- Recombinant proteins with no detectable protease contaminating activities