Order Entry
Puerto Rico
ContactUsLinkComponent
Hi-T7 RNA Polymerase (High Concentration), New England Biolabs
Hi-T7 RNA Polymerase (High Concentration), New England Biolabs
Catalog # 76407-350
CAS Number:  
undefined
Hi-T7 RNA Polymerase (High Concentration), New England Biolabs
Catalog # 76407-350
Supplier Number:  M0470T
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Hi-T7 RNA Polymerase (high concentration)
  • Size:
    50000 units
  • Cat. No.:
    76407-350

Specifications

About this item

Hi-T7 RNA Polymerase (High Concentration) is offered at a 20-fold higher concentration than our standard Hi-T7 RNA Polymerase and is ideal for experienced users interested in building and optimizing their own in vitro transcription reactions.

  • Active from 37 to 56 °C, optimal incubation temperature is 50 to 52 °C
  • In vitro transcription using T7 phage promoter
  • High concentration version of Hi-T7 RNA Polymerase

The enzyme is accompanied by Hi-T7 RNA Polymerase reaction buffer and Magnesium chloride solution to further enable testing of different conditions such as nucleotide concentrations. In vitro transcription involves multiple components in addition to the RNA Polymerase including ribonucleotides, inorganic pyrophosphatase and RNase inhibitor, which can be purchased separately. Please refer to the Related Products section for details. Hi-T7 RNA Polymerase is an engineered DNA-dependent RNA polymerase that is highly specific for T7 phage promoters and is designed to function at higher temperatures than the wild-type bacteriophage T7 RNA Polymerase.

Concentration = 1000000 units/ml.