Scotch-Weld™ Epoxy Adhesive DP190, Gray, 48.5 ml Duo-Pak, 3M™
Supplier: 3M
Certificates
About this item
3M™ Scotch-Weld™ Epoxy Adhesive DP190 is a two-part epoxy adhesive that delivers exceptional performance with high shear and peel strength. Extremely compatible, we formulated this epoxy adhesive to bond to a wide range of materials, such as metals, ceramics, wood, fiberboard, glass, rubber and many plastics.
- Use a tough high shear and peel strength adhesive
- Apply with ease with a low viscosity formulation
- Get a two-part epoxy adhesive that’s highly compatible with many different materials
- Formulated with low viscosity for easy dispensing
Bond 3M™ Scotch-Weld™ Epoxy Adhesive DP190 to a wide range of materials, such as metals, ceramics, wood, fibreboard, glass, rubber and many plastics. Our two-part epoxy adhesive has high shear and peel strength. It adheres with tough, strong bonds and offers an extended work life. The adhesive is designed with low viscosity formulation to give additional time for adjustment and make application easy.
Packaging: 12/case.
Specifications
- Catalog No:
- 76439-340
- 76439-342
- Viscosity:
- Self-Leveling
- Self-Leveling
- Temperature Range:
- 16…27 °C
- 16…27 °C
- Resistance:
- UL94 HB
- UL94 HB