Specifications
- Antibody Type:Primary
- Antigen Name:Green Fluorescent Protein (GFP)
- Antigen Symbol:GFP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western Blot:Yes
- Size:100 µg
- Form:Lyophilized
- Gene ID:Uniprot# Q6YGZ0
- Shipping Temperature:RT
- Immunogen:Recombinant AcGFP purified from E. coli
- Purification:Affinity-purified
- Cat. No.:76459-468
Specifications
About this item
The green fluorescent protein (GFP) is a 27 kDa protein isolated originally from the jellyfish Aequoria victoria. It has an endogenous fluorochrome activity with excitation maximum at 395nm and emission maximum at 509 nm, which is similar to that of fluorescein. GFP can be expressed in fluorescent form in essentially any prokaryotic or eukaryotic cell.
This GFP rabbit antibody was made against a recombinant GFP construct originating from an Aequoria species which was engineered to improve spectral properties and prevent oligomerization (1). This form of GFP, referred to as AcGFP, is 94% identical to the eGFP developed by Tsien and co-workers. The antibody can be used to verify the expression, size and stability of both AcGFP and eGFP fusion proteins in western blotting experiments and to amplify GFP signals in tissues of transgenic animals.
Applications: This antibody is recommended for use in immunocytochemistry (1:2,000 - 1:5,000) and western blotting (1:1,000 - 1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Species independent