Order Entry
Puerto Rico
ContactUsLinkComponent
StickTogether DNA Ligase Buffer, New England Biolabs
StickTogether DNA Ligase Buffer, New England Biolabs
Catalog # 76423-398
CAS Number:  
undefined
StickTogether DNA Ligase Buffer, New England Biolabs
Catalog # 76423-398
Supplier Number:  B0535S
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    2X
  • Description:
    StickTogether DNA ligase buffer
  • Size:
    2 ml
  • Cat. No.:
    76423-398

Specifications

About this item

StickTogether Buffer is supplied with T3, T7, Hi-T4 and Salt-T4 DNA ligases.

  • At a 1X concentration, this reaction buffer ensures optimal activity of these DNA ligases
  • Provided as a 2X solutionContains 2 vials of buffer

New England Biolabs supplies a reaction buffer with all of its enzymes.