Order Entry
United States
ContactUsLinkComponent
 
Human PLAT/TPA PicoKine; Quick ELISA Kit, Boster
Catalog # 76467-400
CAS Number:  
undefined
Human PLAT/TPA PicoKine; Quick ELISA Kit, Boster
Catalog # 76467-400
Supplier Number:  FEK0897
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic box
  • Assay Type (ELISA with LOV):
    Sandwich
  • Host:
  • Primary antibody reactivity:
    Human
  • Size:
    96 wells/kit, with removable strips
  • Environmentally Preferable:
  • Storage Conditions:
    Avoid multiple freeze−thaw cycles (Shipped with wet ice.)
  • Storage Temperature:
    Store at 4 °C for 6 months, at −20 °C for 12 months.
  • Cat. No.:
    76467-400

Specifications

About this item

The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detect Human Tpa/PLAT with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernatants, serum, plasma (heparin, EDTA, citrate) and urine. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Tpa/PLAT tissue specificity: Synthesized in numerous tissues (including tumors) and secreted into most extracellular body fluids, such as plasma, uterine fluid, saliva, gingival crevicular fluid, tears, seminal fluid, and milk.