Mouse IL-23R ELISA Kit EZ-Set (DIY Antibody Pairs), Boster
Catalog # 76467-284
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Pack type:Plastic box
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Target protein:IL-23
- Size:For 5 plates, 96 wells each
- Storage Conditions:Avoid multiple freeze−thaw cycles (Shipped with wet ice.)
- Storage Temperature:Store at 4 °C for 6 months, at −20 °C for 12 months.
- Cat. No.:76467-284
Specifications
About this item
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects IL-23R in Mouse samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.