Human KLK1 ELISA Kit EZ-Set (DIY Antibody Pairs), Boster
Catalog # 76467-232
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Pack type:Plastic box
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Size:For 5 plates, 96 wells each
- Storage Conditions:Avoid multiple freeze−thaw cycles (Shipped with wet ice.)
- Storage Temperature:Store at 4 °C for 6 months, at −20 °C for 12 months.
- Cat. No.:76467-232
Specifications
About this item
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects KLK1 in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC.