Anti-MUT Mouse Monoclonal Antibody [clone: 2D6]
Catalog # 76467-722
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:A synthetic peptide corresponding to a sequence at the N-terminus of human MUT (33-65aa LHQQQPLHPEWAALAKKQLKGKNPEDLIWHTPE), different from the related mouse sequence by one amino acid.
- Antigen Symbol:MUT
- Clonality:Monoclonal
- Clone:2D6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ug/vial
- Form:Lyophilized
- Storage Temperature:At -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquoted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human MUT (33-65aa LHQQQPLHPEWAALAKKQLKGKNPEDLIWHTPE), different from the related mouse sequence by one amino acid.
- Purification:Immunogen affinity purified.
- Cat. No.:76467-722
Specifications
About this item
Mouse IgG monoclonal antibody for Methylmalonyl Coenzyme A mutase detection. Tested with WB, IHC-P in Human,Mouse,Rat.
Type: Primary
Antigen: MUT
Clonality: Monoclonal
Clone: 2D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human,Mouse,Rat