Anti-cavin4a Rabbit Polyclonal Antibody
Catalog # 76466-264
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Symbol:cavin4a
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish
- Size:200 ul/vial
- Form:Liquid
- Storage Temperature:At -20°C for one year, at 4°C for one month. Avoid repeated freezing and thawing.
- Concentration:500μg/ml
- Purification:Immunogen affinity purified.
- Cat. No.:76466-264
Specifications
About this item
Rabbit IgG polyclonal antibody for Cavin4a detection.
Type: Primary
Antigen: cavin4a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Zebrafish