Anti-NPC2 Rabbit Polyclonal Antibody (For Niemann Pick C2/NPC2 detection)
Catalog # 76464-478
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:E.coli-derived human Niemann Pick C2/NPC2 recombinant protein (Position: E20-L151).
- Antigen Symbol:NPC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ug/vial
- Form:Lyophilized
- Storage Temperature:At -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquoted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
- Immunogen:E.coli-derived human Niemann Pick C2/NPC2 recombinant protein (Position: E20-L151).
- Purification:Immunogen affinity purified.
- Cat. No.:76464-478
Specifications
About this item
Rabbit IgG polyclonal antibody for Niemann Pick C2/NPC2 detection. Tested with WB, IHC-P, ICC/IF, FCM, Direct ELISA in Human,Mouse,Rat.
Type: Primary
Antigen: NPC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,Mouse,Rat