Anti-PRKAA1 Rabbit Polyclonal Antibody (For AMPK alpha 1/PRKAA1 detection)
Catalog # 76464-062
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:E.coli-derived human AMPK alpha 1/PRKAA1 recombinant protein (Position: D359-P539).
- Antigen Symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- Reactivity:Monkey,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ug/vial
- Form:Lyophilized
- Storage Temperature:At -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquoted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
- Concentration:Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
- Immunogen:E.coli-derived human AMPK alpha 1/PRKAA1 recombinant protein (Position: D359-P539).
- Purification:Immunogen affinity purified.
- Cat. No.:76464-062
Specifications
About this item
Rabbit IgG polyclonal antibody for AMPK alpha 1/PRKAA1 detection. Tested with WB, FCM, Direct ELISA in Human,Mouse,Rat,Monkey.
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,Monkey,Mouse,Rat