Anti-PRKAA1 Rabbit Polyclonal Antibody (For AMPK alpha 1 detection)
76464-060
- Antibody Type:Primary
- Antigen Name:E. coli-derived human AMPK alpha 1 recombinant protein (Position: F377-R446).
- Antigen Symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ug/vial
- Form:Lyophilized
- Storage Temperature:At -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquoted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
- Concentration:0.5-1mg/ml, actual concentration vary by lot. Use suggested dilution ratio to decide dilution procedure.
- Immunogen:E. coli-derived human AMPK alpha 1 recombinant protein (Position: F377-R446).
- Cat. No.:76464-060
Rabbit IgG polyclonal antibody for AMPK alpha 1 detection. Tested with WB, ICC/IF, FCM, Direct ELISA in Human,Mouse,Rat.
Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,Mouse,Rat