Order Entry
United States
ContactUsLinkComponent
Luna® Cell Ready Probe One-Step RT-qPCR Kit, New England Biolabs
Luna® Cell Ready Probe One-Step RT-qPCR Kit, New England Biolabs
Catalog # 76407-358
CAS Number:  
undefined
Luna® Cell Ready Probe One-Step RT-qPCR Kit, New England Biolabs
Catalog # 76407-358
Supplier Number:  E3031S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
    RT-qPCR probe
  • One step:
    Yes
  • Description:
    Luna® Cell Ready Probe One-Step RT-qPCR kit
  • Size:
    100 reactions
  • Environmentally Preferable:
  • Cat. No.:
    76407-358

Specifications

About this item

The Luna® Cell Ready Probe One-Step RT-qPCR Kit provides all the necessary components for direct, probe-based RNA detection and quantitation, bypassing the need for RNA extraction and purification.

  • Contains reagents for 100 lysis preps and 500 one-step RT-qPCR reactions (probe)
  • Effective cell lysis preparation from 10 to 100000 cells across numerous cell lines
  • Coordinated cell lysis, RNA release, and genomic DNA removal in a fast 15-minute protocol
  • Luna WarmStart RT paired with Hot Start Taq for increased thermostability and room temperature setup

Coordinating the actions of DNase I and the Luna Cell Ready Protease, the Luna Cell Ready Lysis Module offers a simple alternative workflow resulting in effective cell lysis, RNA release, and genomic DNA removal simultaneously in a 15-minute protocol. The Lysis Module includes a unique Luna Cell Ready RNA Protection Reagent that maintains RNA integrity during cell lysis. The lysis capacity spans 10 to 100000 cells in a 50 µl lysis reaction. Up to 2 µl of lysate (equivalent to RNA from 0.2 to 4000 cells) can be transferred into 20 µl downstream RT-qPCR reactions.

Delivery: The Luna Cell Ready Probe One-Step RT-qPCR Kit is composed of:
1) The Luna Cell Ready Lysis Module
2) The Luna Universal Probe One-Step RT-qPCR Kit.