ContactUsLinkComponent
SHuffle T7 Express Competent E. coli, New England Biolabs
SHuffle T7 Express Competent E. coli, New England Biolabs
  103391-806
undefined
SHuffle T7 Express Competent E. coli, New England Biolabs
  103391-806
 :  C3029J
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    SHuffle T7 Express competent E.coli
  • Size:
    12 vials×0.05 ml/tube
  • Cat. No.:
    103391-806
Chemically competent E. coli cells suitable for T7 protein expression with enhanced capacity to correctly fold proteins with multiple disulfide bonds in the cytoplasm.
  • T7 expression
  • Engineered E. coli B strain to promote disulfide bond formation in the cytoplasm
  • Deficient in proteases Lon and OmpT
  • Resistant to phage T1 (fhuA2)
  • Free of animal products