Order Entry
Puerto Rico
ContactUsLinkComponent
 
Flammable
Toxic
Health
Methanol Dehydrant, Epredia™ for histology
Catalog # 72060-080
Supplier:  Epredia
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol Dehydrant, Epredia™ for histology
Catalog # 72060-080
Supplier:  Epredia
Supplier Number:  9611
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Poly bottle
  • Size:
    1 gal.
  • Cat. No.:
    72060-080
  • Supplier no.:
    9611

Specifications

About this item

 :
 :32.04 g/mol
Flash Pt:11 °C
Storage Temperature:Ambient
UN:1230
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595