Mouse Artemin PicoKine; ELISA Kit, Boster
76466-398
:
- Pack type:Plastic box
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Size:96 wells/kit, with removable strips
- Storage Conditions:Avoid multiple freeze−thaw cycles (Shipped with wet ice.)
- Storage Temperature:Store at 4 °C for 6 months, at −20 °C for 12 months.
- Cat. No.:76466-398
Sandwich high sensitivity ELISA kit for quantitative detection of Mouse Artemin. 96 wells/kit, with removable strips.