Orders LinkContactUsLinkComponent
Everlast™ 247 Rocker, Electron Microscopy Sciences
Everlast™ 247 Rocker, Electron Microscopy Sciences
247 rocker
Catalog # 76421-792
Supplier #:  65700-09
Everlast™ 247 Rocker, Electron Microscopy Sciences
Catalog # 76421-792
Everlast™ 247 Rocker, Electron Microscopy Sciences
Everlast™ 247 Rocker, Electron Microscopy Sciences
247 rocker
Price
$2,474.02
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Everlast™ 247 Rocker is a powerful motor and heavy-duty drive system.
  • Extra large 16×12" work surface
  • Optional stacking platform doubles the working area
  • Platforms made of stainless steel to prevent corrosion or rusting
  • Provides years of uninterrupted service
They are consistent and reliable, even with constant use in cold rooms or incubators. The rocking speed and tilt angle are easily adjustable for gentle or aggressive agitation.
Specifications
  • Description:
    247 rocker
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Stacking platform with flat mat for Everlast
    Stacking platform with flat mat for Everlast
    76421-794
    Log in for availability
    Each - $302.98