Order Entry
Puerto Rico
ContactUsLinkComponent
Monarch® Protein Separation Solution, New England Biolabs
Monarch® Protein Separation Solution, New England Biolabs
Catalog # 76470-774
CAS Number:  
undefined
Monarch® Protein Separation Solution, New England Biolabs
Catalog # 76470-774
Supplier Number:  T3062L
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Monarch® protein separation solution
  • Cat. No.:
    76470-774
  • Volume:
    36 ml

Specifications

About this item

The Monarch® Protein separation solution, supplied as a component of the Monarch HMW DNA extraction kit for tissue, is a salt-based solution that is optimized to ensure proteins in tissue and bacteria samples are easily removed during the phase separation of the Monarch HMW DNA extraction workflow.

  • Facilitates the separation of proteins during HMW DNA extraction from tissues
  • Salt-based solution for protein precipitation
Delivery: Supplied in 2 bottles of 18 ml.