Specifications
- Description:Monarch® gDNA nuclei Prep and lysis buffer pack
- Size:1 pack
- Cat. No.:76470-766
Specifications
About this item
The Monarch® gDNA Nuclei Prep and lysis buffer pack contains two lysis buffers used in the Monarch HMW DNA extraction kit for cells and blood.
- Contains Monarch® gDNA Nuclei Prep buffer and Monarch gDNA Nuclei lysis buffer
- Supplied as components of the Monarch HMW DNA Extraction kit for cells and blood
- Used for rapid and efficient lysis of mammalian cells
- 2-step lysis enables highly effective RNA removal
- 20 ml of each buffer is included in the buffer pack
The Nuclei Prep Buffer is a mild lysis buffer used first to lyse the cell membrane and release the contents of the cytoplasm while the nuclei are kept intact. It is used pre-mixed with the RNase A enzyme included in the kit. After the RNA from the cytoplasm is digested by RNase A, the Nuclei Lysis Buffer, pre-mixed with the included Proteinase K, is used to lyse the nuclei and release the HMW genomic DNA. This two-step lysis ensures that RNA in the cytoplasm is effectively degraded before the viscous genomic DNA from the nuclei is released.
The Nuclei Prep Buffer is used chilled to 4 °C, and it can be stored at 4 °C for convenience if desired.