Specifications
- Description:Monarch® bead retainers
- Size:100 pieces
- Cat. No.:76470-764
Specifications
About this item
Plastic parts for retaining the Monarch DNA capture beads during the Monarch HMW DNA extraction workflow. These pieces resemble spin columns without any membrane inserted and facilitate various steps of the extraction workflow. They mate to the Monarch collection tube II.
- Supplied as a component of the Monarch HMW DNA extraction kits
- Facilitates Monarch HMW DNA extraction workflow by containing the beads during wash and elution
- Plastic part resembling a spin column without a membrane