ContactUsLinkComponent
Monarch® Tubes, 2 ml, New England Biolabs
  76470-760
undefined
Monarch® Tubes, 2 ml, New England Biolabs
  76470-760
 :  T3003L
  • Capacity:
    2 ml
  • Description:
    Monarch® tubes, 100 tubes/pack
  • Cat. No.:
    76470-760
The Monarch® tubes are supplied as a component of the Monarch HMW DNA extraction kits.
  • Includes locking mechanism on the cap
The tubes are round-bottom and have a locking mechanism for safe and convenient handling. The tubes are low DNA binding, making them optimal for purification of HMW DNA in the Monarch workflows.