Monarch® Pestle Set, New England Biolabs
76470-758
- Description:Monarch pestle set
- Size:100 sets
- Cat. No.:76470-758
The Monarch® pestle set contains single-use microtube pestles and the accompanying 1.5 ml tubes for sample disruption and homogenization prior to nucleic acid extraction. 100 green pestles and tubes are included in this set.
- Set includes Monarch pestles and pestle tubes
- Optimally designed for manual homogenization of tissue samples prior to nucleic acid extraction
- Single-use pestles and tubes