SP Bel-Art® Sterileware® Single Slot Powder Sampler, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Ideal for taking a single, targeted 50 ml sample from free-flowing powders and granules.
- After insertion, rotate tube to open then close the collection slot, and remove to retrieve the sample
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) sampler heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10⁻⁶)
Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization.