jetOPTIMUS® Buffer, Polyplus-transfection
jetOPTIMUS® buffer, 60 ml
Catalog # 76327-128
Supplier: Polyplus Transfection
Supplier #: 201000001
jetOPTIMUS® Buffer, Polyplus-transfection
Catalog # 76327-128
Supplier: Polyplus Transfection
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Dedicated buffer for jetOPTIMUS®, best-in-class DNA transfection reagent.
Certifications: Since 2002, Polyplus-transfection® is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Specifications
Specifications
- Description:jetOPTIMUS® buffer
- Buffer volume:60 ml



