Order Entry
Puerto Rico
ContactUsLinkComponent
Phosphate Buffered Saline (10X PBS), pH 7.2, Sterile, Quality Biological
Phosphate Buffered Saline (10X PBS), pH 7.2, Sterile, Quality Biological
Catalog # 76470-186
   Sustainable Packaging
CAS Number:  
undefined
Phosphate Buffered Saline (10X PBS), pH 7.2, Sterile, Quality Biological
Catalog # 76470-186
Supplier Number:  119-248-161
   Sustainable Packaging
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    PBS buffer (10X), pH 7.2
  • Size:
    20 L
  • Environmentally Preferable:
    Sustainable Packaging
  • Shelf Life:
    24 months
  • Cat. No.:
    76470-186

Specifications

About this item

PBS (phosphate buffered saline) is a balanced salt solution used for a variety biological and cell culture applications.

  • Use for biological, cell-culture, and immunological applications
  • Sterile filtered
  • Endotoxin and Mycoplasma tested

PBS used for a variety of applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. It is also useful in immunological applications, such as Western Blotting.