Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Plier, Roll Pin Insertion-Extraction, Excelta

Plier, Roll Pin Insertion-Extraction, Excelta

Pliers, roll pin extraction, 5 star quality, Carbon, 162.50 mm (6.5")
Catalog # 63043-006
Supplier:   Excelta
Supplier #:  900-100A
Plier, Roll Pin Insertion-Extraction, Excelta
Catalog # 63043-006
Supplier:  Excelta
Plier, Roll Pin Insertion-Extraction, Excelta
Plier, Roll Pin Insertion-Extraction, Excelta

Pliers, roll pin extraction, 5 star quality, Carbon, 162.50 mm (6.5")

Price
$1,151.80
Log in for availability
Each
Product Details & Documents

The 900-100 series are designed to remove or add the card key roll pin.

  • Roll pin insertion, extraction tool
  • Replaceable pin
  • Ergonomic handles, dual leaf springs

These 6.5" roll pin insertion/extraction pliers are designed to insert/extract the card key roll pin, thus reducing production time. The 0.046" pin is replaceable. They features soft cushioned ergonomic grips (10¹⁰ ohms/sq surface resistivity) with dual leaf springs for smooth action. All are made of carbon steel.

Specifications
  • Description:
    Pliers, roll pin extraction, 5 star quality
  • Length:
    162.50 mm (6.5")
  • Material:
    Carbon
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.