Orders LinkContactUsLinkComponent
Wire Stripper, Excelta
Wire Stripper, Excelta
Supplier:   Excelta
Wire Stripper, Excelta
Supplier:  Excelta
Certificates
Wire Stripper, Excelta
Wire Stripper, Excelta
1 Options Available Below
Product Details & Documents
These two tools are designed for stripping PTFE coated small wires.
  • Will strip PTFE coating in one shot
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use
These tools are made to strip small PTFE coated wire up to 30AWG. The 502-US will strip one size wire; (specify the wire gauge size by indicating the gauge at the end of the reference number. Available in 22, 24, 26, 28, and 30 AWG.) The 510E-US is a 'V' notched wire stripper which cuts the insulation on four sizes makeing stripping much easier. It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. Both are made of carbon tool steel with flame hardened edges.
Product Family Options
Product Information
  • Material
    Length
    Availability
    Price
  • 63042-432
    Pliers, wire stripper, 5 star quality, custom size wire
    Pliers, wire stripper, 5 star quality, custom size wire
    Carbon
    137.5 mm (5.5")
    Discontinued
    Supplier Number: 502E-US
  • 63042-500
    Pliers, wire stripper, v-notch, 5 star quality, adjustable
    Pliers, wire stripper, v-notch, 5 star quality, adjustable
    Carbon
    137.5 mm (5.5")
    Log in for availability
    Log in for availability
    Each
    Each - $2,914.63
    $2,914.63
    Each
       $2,914.63
    Supplier Number: 510E-US