Order Entry
Puerto Rico
ContactUsLinkComponent
Brady® BradyPrinter i7100 300 DPI Peel Model with Vial Applicator Kit, Brady Worldwide
Brady® BradyPrinter i7100 300 DPI Peel Model with Vial Applicator Kit, Brady Worldwide
Catalog # 76461-896
Supplier:  Brady Worldwide
undefined
Brady® BradyPrinter i7100 300 DPI Peel Model with Vial Applicator Kit, Brady Worldwide
Catalog # 76461-896
Supplier:  Brady Worldwide
Supplier Number:  151752

Specifications

  • Description:
    Single color printer
  • Cat. No.:
    76461-896
  • Resolution:
    300 dpi
  • Dimensions:
    11.4H×9.9W×17.2" D
  • Software:
    Brady Workstation; Windows based driver for 3rd party software use
  • Label Size:
    2.2" max. label/tape width

Specifications

About this item

Get everything you need to automate your vial identification in one complete kit with the BradyPrinter i7100 Vial Applicator Kit.

  • Color touch screen
  • 256 MB RAM; SD Flash memory card reader
  • PC Connect mode of operation

Get the job done with the BradyPrinter i7100 label printer. No matter the demands, this reliable, heavy-duty and high-accuracy label printer gets it all done. BradyPrinter i7100 Vial Applicator easily applies to vials during your high-volume identification projects. Save up to 5% when you buy the kits vs. buying each piece individually. Using printed labels allow for precision tracking by eliminating error-prone, handwritten labels. Vial applicator pays for itself after approximately 100 hours of use.

Delivery: Includes (1) i7100 300 dpi Peel Model Printer (149045); (1) Vial Label Applicator; (1) Brady Workstation Product and Wire ID Software Suite (BWS-PWID-EM); (1) Configuration Settings Manual; (1) Owners Manual; (1) Product CD with Driver; (1) U.S. Type B Plug Power Cord; (1) USB Cable; (2) 1 In Ribbon Takeup Cores.