Orders LinkContactUsLinkComponent
Removable Polyester Labels, Brady
Removable Polyester Labels, Brady
Supplier:   Brady Worldwide
Removable Polyester Labels, Brady
Supplier:  Brady Worldwide
Certificates
Removable Polyester Labels, Brady
Removable Polyester Labels, Brady
6 Options Available Below
Product Details & Documents
Removable Polyester (B-522) is ideal for alphanumeric and barcode applications such as electronic component marking and general purpose applications.
  • Gloss finish
  • Removable acrylic adhesive type
Polyester with removable adhesive is thermal transfer printable. Good solvent resistance, heat resistance and clean removability. Service temperature of −94 to 93 °C (−70 to 200 °F). If using this part in an IP series printer or i5100 printer, white print ribbon is not recommended.
Product Family Options
Product Information
  • Material
    Color
    For Use With
    Core size
    Dimensions
    Packaging
    Availability
    Price
  • 76461-906
    Removable polyester labels
    Removable polyester labels
    Polyester
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    11.1×12.7 mm (0.44×0.5")
    10000 labels/roll, 1 roll (10000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $220.66
    $220.66
    Roll of 1
       $220.66
    Supplier Number: THT-12-522-10
  • 76461-916
    Removable polyester labels
    Removable polyester labels
    Polyester
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    12.7×25.4 mm (0.5×1")
    20000 labels/roll, 1 rolls (20000 labels)
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $304.98
    $304.98
    Pack of 2
       $304.98
    Supplier Number: THT-59-522-10
  • 76461-914
    Removable polyester labels
    Removable polyester labels
    Polyester
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    12.7×25.4 mm (0.5×1")
    1000 labels/roll, 2 rolls (20000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $360.11
    $360.11
    Roll of 1
       $360.11
    Supplier Number: THT-5-522-10
  • 76461-902
    Removable polyester labels
    Removable polyester labels
    Polyester
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    25.4×50.8 mm (1×2")
    3000 labels/roll, 1 roll (3000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $282.25
    $282.25
    Roll of 1
       $282.25
    Supplier Number: THT-17-522-3
  • 76461-922
    Removable polyester labels
    Removable polyester labels
    Polyester
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    25.4×76.2 mm (1×3")
    3000 labels/roll, 1 roll (3000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $450.96
    $450.96
    Roll of 1
       $450.96
    Supplier Number: THT-18-522-3
  • 76461-924
    Removable polyester labels
    Removable polyester labels
    Polyester
    Clear
    BradyPrinter i5100 industrial label printer; BradyPrinter i7100 industrial label printer; THT 3" Core printers
    3" Ø
    50.8×76.2 mm (2×3")
    1000 labels/roll, 1 roll (1000 labels)
    Log in for availability
    Log in for availability
    Roll of 1
    Roll of 1 - $301.70
    $301.70
    Roll of 1
       $301.70
    Supplier Number: THT-19-522-1