Orders LinkContactUsLinkComponent
Immunocard® Mycoplasma, Meridian Bioscience
Immunocard® Mycoplasma, A rapid, sensitive enzyme immunoassay for the detection of IgM to Mycoplasma pneumoniae in human serum
Catalog # 76437-438
Supplier:   Meridian
 :  709030
Immunocard® Mycoplasma, Meridian Bioscience
Catalog # 76437-438
Supplier:  Meridian
CAS Number:  
Immunocard® Mycoplasma, Meridian Bioscience
Immunocard® Mycoplasma, Meridian Bioscience
Immunocard® Mycoplasma, A rapid, sensitive enzyme immunoassay for the detection of IgM to Mycoplasma pneumoniae in human serum
CAS Number:  
Price
$1,628.62
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The ImmunoCard Mycoplasma enzyme immunoassay (EIA) is an in vitro qualitative procedure for the detection of IgM to Mycoplasma pneumoniae in human serum. Test results are intended to aid in the diagnosis of recent Mycoplasma pneumoniae infection.
Specifications
Specifications
  • Description:
    Immunocard® Mycoplasma
  • Details:
    A rapid, sensitive enzyme immunoassay for the detection of IgM to Mycoplasma pneumoniae in human serum
Specifications