ContactUsLinkComponent
Immunocard STAT!® MONO, Meridian Bioscience
Catalog # 76437-440
Supplier:  Meridian
CAS Number:  
undefined
Immunocard STAT!® MONO, Meridian Bioscience
Catalog # 76437-440
Supplier:  Meridian
Supplier Number:  755725
CAS Number:  
Specifications
  • Description:
    Immunocard STAT!® MONO
  • Cat. No.:
    76437-440
  • Details:
    A CLIA waived one step lateral flow immunoassay for the detection of infectious mononucleosis from serum, plasma or whole blood specimens
Specifications
About this item
The Immunocard STAT!® Mono Test is intended for the qualitative detection of infectious mononucleosis heterophile antibodies in serum, plasma or whole blood as an aid in the diagnosis of infectious mononucleosis.
The ImmunoCard STAT! Mono Test uses color immunochromatographic dipstick technology with bovine erythrocyte extract coated on the membrane. In the test procedure, serum, plasma or whole blood is mixed with the Diluent. Then the Test Stick is placed in the mixture and the mixture migrates along the membrane. If the specific IM heterophile antibody is present in the sample, it will form a complex with the bovine erythrocyte extract conjugated color particles. The complex will then be bound by bovine erythrocyte extract immobilized on the membrane and a visible blue Test Line will appear to indicate a positive result.