Immunocard STAT!® Strep A, Meridian
Immunocard STAT!® Strep A
Catalog # 76437-434
Supplier: Meridian
Supplier #: 755250
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Alethia Group A Streptococcus assay, is a qualitative invitro diagnostic test for the detection of Streptococcus pyogenes (Group A β-hemolytic Streptococcus) in throat swab specimens.
- CLIA Waived, easy to use technology
- Rapid test for detection of Group A Streptococcal Antigen from throat swabs
- Results in five minutes, shortening patient wait time
- Test with Immunocard STAT! Strep A first and be able to treat while patient is waiting
- Reflex Rapid Group A Strep negatives to the Alethia Group A Strep molecular to confirm results
Specifications
Specifications
- Description:Immunocard STAT!® Strep A