Orders LinkContactUsLinkComponent
Alethia® <i>C. difficile</i> DNA amplification assay, Meridian Bioscience
Alethia® C. difficile DNA amplification assay, Meridian Bioscience
Alethia® C. difficile, Molecular assay for the detection of C. difficile. Alethia® provides a quick, accurate and reliable molecular assay for C. difficile detection.
Catalog # 76437-450
Supplier:   Meridian
Supplier #:  480050
Alethia® <i>C. difficile</i> DNA amplification assay, Meridian Bioscience
Catalog # 76437-450
Supplier:  Meridian
CAS Number:  
Alethia® <i>C. difficile</i> DNA amplification assay, Meridian Bioscience
Alethia® C. difficile DNA amplification assay, Meridian Bioscience
Alethia® C. difficile, Molecular assay for the detection of C. difficile. Alethia® provides a quick, accurate and reliable molecular assay for C. difficile detection.
CAS Number:  
Price
$7,015.47
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The Alethia C. difficile DNA amplification assay, performed on the Alethia reader, is a qualitative in-vitro diagnostic test for the direct detection of toxigenic C. difficile in human stool specimens from pediatric and adult patients suspected of having Clostridium difficile-associated disease (CDAD).
  • Reduce the risk of missing a true positive with the accuracy of molecular
Fast, actionable results enable physicians to make informed decisions about the patient management and treatment resulting in reduced healthcare costs associated with isolation and antibiotics.
Specifications
Specifications
  • Description:
    Alethia® C. difficile
  • Details:
    Molecular assay for the detection of <i>C. difficile</i>. Alethia® provides a quick, accurate and reliable molecular assay for C. difficile detection.