Alethia® Group A Streptococcus, Meridian Bioscience
Catalog # 76437-452
Supplier: Meridian
CAS Number:
Specifications
- Description:Alethia® Group A Streptococcus
- Cat. No.:76437-452
- Details:Molecular assay for the detection of Group A Streptococcus. A quick, accurate and reliable molecular assay for Group A Strep detection.
Specifications
About this item
The Alethia Group A Streptococcus (Group A Strep) assay, performed on the Alethia reader is a qualitative in vitro diagnostic test for the detection of Streptococcus pyogenes (Group A β-hemolytic Streptococcus) in throat swab specimens.
- The accuracy and speed of Alethia Group A Strep results helps families return to their busy daily routines as quickly as possible
- Alethia Group A Strep molecular amplification has a 98% sensitivity that has shown to be 53% more sensitive than traditional culture
Alethia Group A Strep can help prevent unncessary antibiotic treatment which can lead to the development of antimicrobial resistance among common pathogens.