Alethia® Group B Streptococcus, Meridian Bioscience
Catalog # 76437-456
Supplier: Meridian
CAS Number:
Specifications
- Description:Alethia® Group B Streptococcus
- Cat. No.:76437-456
- Details:Molecular assay for the detection of Group B Streptococcus.
Specifications
About this item
The Alethia® Group B Streptococcus assay is a qualitative in vitro diagnostic for the detection of Streptococcus agalactiae in enriched cultures obtained from vaginal/rectal swab specimens from antepartum women.