Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant CD30 (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)
Human Recombinant CD30 (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)
Certificates
Human Recombinant CD30 (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)

Specifications

  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Protein Synonyms:
    Ki-1|CD30|D1S166E|TNFRSF8
  • Protein/Peptide Name:
    CD30
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    64.6 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Please contact us for detailed information.
  • Shipping Temperature:
    RT

Specifications