Human Recombinant CD27 Ligand (from HEK293 Cells)
Human CD27 Ligand / CD70 Protein, Fc Tag (active trimer) (MALS verified), ACROBiosystems
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag (active trimer) (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human CD27, His Tag (Cat. No. CD7-H522b) at 2 μg/mL (100 μL/well) can bind Human CD27 Ligand, Fc Tag (active trimer) (MALS verified) (Cat. No. CDL-H5266) with a linear range of 0.2-8 ng/mL (QC tested).
- Gene ID:P32970-1
- Protein Synonyms:CD27LG|TNFSF7|TNFSF7G|CD27L|CD70
- Protein/Peptide Name:CD27 Ligand
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:74.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris, Glycine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT