Human Recombinant CD23 (from HEK293 cells)
103814-842
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:50 µg
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P06734-1
- Protein Synonyms:CLEC4J|FCE2|CD23A|CD23|IGEBF|FCER2|FcεRII|FcεRⅡ
- Protein/Peptide Name:CD23
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:32.9 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
- Cat. No.:103814-842
Human CD23 / Fc epsilon RII Protein, His Tag, ACROBiosystems