Human Recombinant BTLA (from HEK293 cells)
Human BTLA (31-150) Protein, Mouse IgG2a Fc Tag, ACROBiosystems
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Gene ID:Q7Z6A9-1(V105M,S138G,M148V)
- Protein Synonyms:BTLA|CD272
- Protein/Peptide Name:BTLA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:40.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 100 mM Glycine, 150 mM NaCl, Arginine, pH3.2. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT