Human Recombinant BAFF (from HEK293 cells)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Human BAFF / TNFSF13B / CD257 Protein, His,Flag Tag, active trimer (MALS verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:active trimer (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Human BAFFR Protein, Fc Tag (Cat. No. BAR-H5257) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human BAFF Protein, His,Flag Tag (Cat. No. BAF-H52D4) with an affinity constant of 5.54 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Human BAFF, His,Flag Tag, active trimer (Cat. No. BAF-H52D4) stimulates proliferation of mouse spleen cells. The EC50 for this effect is 1.97-2.90 ng/mL (Routinely tested).
- Protein Synonyms:CD257|ZTNF4|THANK|TALL-1|TNFSF20|DTL|TNFSF13B|TALL1|BLYS|BAFF
- Protein/Peptide Name:BAFF
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:52.5 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 25 mM MES, 150 mM NaCl, pH 6.0. Normally Trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT