SARS-CoV-2 Recombinant S2 protein (from HEK293 cells)
SARS-CoV-2 (COVID-19) S2 protein, His Tag, ACROBiosystems
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:SARS-CoV-2
- Storage Conditions:–70 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Protein Synonyms:Spike|S2 protein|S glycoprotein Subunit2|Spike glycoprotein Subunit2|COVID-19|Spike protein S2
- Protein/Peptide Name:S2 protein
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:60.0 kDa
- Endotoxin Level:Low
- Formulation:Delivered as bulk protein in a 0.2 μm filtered solution of 20 mM PB, 300 mM NaCl, pH7.4 with glycerol as protectant.
- Shipping Temperature:Dry Ice
Frequently Bought Together