Order Entry
Puerto Rico
ContactUsLinkComponent
SARS-CoV-2 Recombinant S1 protein (from HEK293 cells)
SARS-CoV-2 Recombinant S1 protein (from HEK293 cells)
Certificates
SARS-CoV-2 Recombinant S1 protein (from HEK293 cells)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Loaded Human ACE2, Fc Tag (Cat. No. AC2-H5257) on Protein A Biosensor, can bind SARS-CoV-2 S1 protein (D614G), His Tag (Cat. No. S1N-C5256) with an affinity constant of 76.4 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Protein Synonyms:
    Spike|Spike glycoprotein Subunit1|S glycoprotein Subunit1|S1 protein|Spike protein S1|COVID-19
  • Protein/Peptide Name:
    S1 protein
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    76.8 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications