SARS-CoV-2 Recombinant Nucleocapsid protein (from E. coli)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
SARS-CoV-2 (COVID-19) Nucleocapsid protein Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Storage Conditions:–70 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Protein Synonyms:NP|Nucleocapsid protein|Protein N|COVID-19
- Protein/Peptide Name:Nucleocapsid protein
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:51.0 kDa
- Endotoxin Level:Low
- Formulation:Delivered as bulk protein in a 0.2 μm filtered solution of 10 mM PB, Arginine, pH7.4.
- Shipping Temperature:Dry Ice