Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Recombinant ACE2 (from HEK293 Cells)
Rat Recombinant ACE2 (from HEK293 Cells)
Certificates
Rat Recombinant ACE2 (from HEK293 Cells)

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Storage Conditions:
    –70 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    Q5EGZ1-1
  • Protein Synonyms:
    ACEH|ACE-2|ACE2
  • Protein/Peptide Name:
    ACE2
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    85.5 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Delivered as bulk protein in a 0.2 μm filtered solution of 50 mM Tris, 150 mM NaCl, Arginine, pH7.5 with glycerol as protectant.
  • Shipping Temperature:
    Dry Ice

Specifications