Paguma larvata Recombinant ACE2 (from HEK293 Cells)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Paguma larvata ACE2 / ACEH Protein, His Tag (SPR verified), ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Paguma larvata
- Tag sequence:His Tag (SPR verified)
- Storage Conditions:–70 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Paguma larvata ACE2, His Tag (Cat. No. AC2-P5248) immobilized on CM5 Chip can bind SARS S protein RBD, His Tag (Cat. No. SPD-S52H6) with an affinity constant of 0.101 μM as determined in a SPR assay (Biacore T200) (QC tested). Paguma larvata ACE2, His Tag (Cat. No. AC2-P5248) immobilized on CM5 Chip can bind SARS-CoV-2 S protein RBD, His Tag (Cat. No. SPD-C52H3) with an affinity constant of 29 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Paguma larvata ACE2, His Tag (Cat. No. AC2-P5248) immobilized on CM5 Chip can bind SARS-CoV-2 S1 protein, His Tag (Cat. No. S1N-C52H4) with an affinity constant of 47.5 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Paguma larvata ACE2, His Tag (Cat. No. AC2-P5248) immobilized on CM5 Chip can bind SARS S1 protein, His Tag (Cat. No. S1N-S52H5) with an affinity constant of 0.796 μM as determined in a SPR assay (Biacore T200) (Routinely tested). Loaded SARS-CoV-2 S protein RBD, Fc Tag (Cat. No. SPD-C5255) on Protein A Biosensor, can bind Paguma larvata ACE2, His Tag (Cat. No. AC2-P5248) with an affinity constant of 1.17 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q56NL1-1
- Protein Synonyms:ACEH|ACE-2|ACE2
- Protein/Peptide Name:ACE2
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:85.7 kDa
- Endotoxin Level:Low
- Formulation:Delivered as bulk protein in a 0.2 μm filtered solution of 50 mM Tris, 150 mM NaCl, pH7.5 with glycerol as protectant.
- Shipping Temperature:Dry Ice