Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant VEGF120 (from HEK293 cells), Biotin
Mouse Recombinant VEGF120 (from HEK293 cells), Biotin
Catalog # 103790-876
CAS Number:  
Mouse Recombinant VEGF120 (from HEK293 cells), Biotin
Catalog # 103790-876
Supplier Number:  VE0M82Q2200UG
CAS Number:  

Specifications

  • Conjugation:
    Biotin
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Size:
    200 µg
  • Tag sequence:
    His, Avitag™
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized VEGFR1/R2 at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His,Avitag (Cat. No. VE0-M82Q2) with a linear range of 0.1-2 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Avastin at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse VEGF120, His,Avitag (Cat. No. VE0-M82Q2) with a linear range of 0.125-2 μg/mL (Routinely tested).
  • Gene ID:
    Q00731-3
  • Protein Synonyms:
    RP1-261G23.1|MVCD1|MGC70609|VEGFA|VPF
  • Protein/Peptide Name:
    VEGF120
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    17.3 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT
  • Cat. No.:
    103790-876

Specifications

About this item

Biotinylated Mouse VEGF120 Protein, His,Avitag™, ACROBiosystems