Mouse Recombinant NOTCH1 (from HEK293 Cells)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Mouse NOTCH1 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:Q01705-1
- Protein Synonyms:NOTCH1|hN1|TAN1|Notch 1
- Protein/Peptide Name:NOTCH1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:55.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT