Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Recombinant CD3E & CD3D (from HEK293 cells)
Mouse Recombinant CD3E & CD3D (from HEK293 cells)
Supplier:  ACRO Biosystems
Certificates
Mouse Recombinant CD3E & CD3D (from HEK293 cells)
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Gene ID:
    P22646-1 (CD3E) & P04235-1 (CD3D)
  • Protein Synonyms:
    CD3 delta / epsilon|CD3 delta & CD3 epsilon|CD3E & CD3D
  • Protein/Peptide Name:
    CD3E & CD3D
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    15.1 kDa (CD3E) & 14.3 kDa (CD3D)
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together