Human Recombinant CD3E & CD3D (from HEK293 Cells)
Human CD3 epsilon & CD3 delta Heterodimer Protein, His Tag&Tag Free (MALS verified), ACROBiosystems
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:His Tag&Tag Free (MALS verified)
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Human CD3E&CD3D Heterodimer Protein, His Tag&Tag Free (Cat. No. CDD-H52W1) captured on CM5 Chip via anti-His antibody can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3) (Cat. No. CDE-M120a) with an affinity constant of 40 nM as determined in SPR assay (Biacore T200) (Routinely tested). Bispecific T-cell Engager (CD3 X BCMA) immobilized on CM5 Chip can bind Human CD3E&CD3D Heterodimer Protein, His Tag&Tag Free (Cat. No. CDD-H52W1) with an affinity constant of 31.8 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
- Gene ID:P07766-1 (CD3E) & P04234-1 (CD3D)
- Protein Synonyms:CD3 delta / epsilon|CD3 delta & CD3 epsilon|CD3E & CD3D
- Protein/Peptide Name:CD3E & CD3D
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:16.6 kDa (CD3E) and 13.4 kDa (CD3D)
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4 . Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Frequently Bought Together